Tisk…

Anti-LENG1 Mouse Monoclonal Antibody [clone: 1E8]

Dodavatel: OriGene
ORIGCF503113EA 21710 CZK
ORIGCF503113
Anti-LENG1 Mouse Monoclonal Antibody [clone: 1E8]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LENG1
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR