Tisk…

Anti-ALDH3A2 Mouse Monoclonal Antibody [clone: 1H10]

Dodavatel: OriGene
ORIGCF503143EA 21120 CZK
ORIGCF503143
Anti-ALDH3A2 Mouse Monoclonal Antibody [clone: 1H10]
Protilátky
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ALDH3A2
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR