Tisk…

Anti-ELK3 Mouse Monoclonal Antibody [clone: 4G9]

Dodavatel: OriGene
ORIGCF503575EA 21120 CZK
ORIGCF503575
Anti-ELK3 Mouse Monoclonal Antibody [clone: 4G9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ELK3
Clonality: Monoclonal
Clone: 4G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR