Tisk…

Anti-PRMT2 Mouse Monoclonal Antibody [clone: 5D10]

Dodavatel: OriGene
ORIGCF503586EA 21710 CZK
ORIGCF503586
Anti-PRMT2 Mouse Monoclonal Antibody [clone: 5D10]
Protilátky
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: PRMT2
Clonality: Monoclonal
Clone: 5D10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR