Tisk…

Anti-CDKN3 Mouse Monoclonal Antibody [clone: 4D8]

Dodavatel: OriGene
ORIGCF503592EA 21120 CZK
ORIGCF503592
Anti-CDKN3 Mouse Monoclonal Antibody [clone: 4D8]
Protilátky
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100

Type: Primary
Antigen: CDKN3
Clonality: Monoclonal
Clone: 4D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR