Tisk…

Anti-SEC14L2 Mouse Monoclonal Antibody [clone: 1B1]

Dodavatel: OriGene
ORIGCF503595EA 21120 CZK
ORIGCF503595
Anti-SEC14L2 Mouse Monoclonal Antibody [clone: 1B1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SEC14L2
Clonality: Monoclonal
Clone: 1B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR