Tisk…

Anti-PSMB9 Mouse Monoclonal Antibody [clone: 1G7]

Dodavatel: OriGene
ORIGCF504350EA 21120 CZK
ORIGCF504350
Anti-PSMB9 Mouse Monoclonal Antibody [clone: 1G7]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PSMB9
Clonality: Monoclonal
Clone: 1G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR