Tisk…

Anti-PSMB7 Mouse Monoclonal Antibody [clone: 1E1]

Dodavatel: OriGene
ORIGCF504368EA 21710 CZK
ORIGCF504368
Anti-PSMB7 Mouse Monoclonal Antibody [clone: 1E1]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: PSMB7
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR