Tisk…

Anti-SERPINA5 Mouse Monoclonal Antibody [clone: 1F8]

Dodavatel: OriGene
ORIGCF504371EA 21120 CZK
ORIGCF504371
Anti-SERPINA5 Mouse Monoclonal Antibody [clone: 1F8]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SERPINA5
Clonality: Monoclonal
Clone: 1F8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR