Tisk…

Anti-PSMB9 Mouse Monoclonal Antibody [clone: 3F3]

Dodavatel: OriGene
ORIGCF504390EA 21120 CZK
ORIGCF504390
Anti-PSMB9 Mouse Monoclonal Antibody [clone: 3F3]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: PSMB9
Clonality: Monoclonal
Clone: 3F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR