Tisk…

Anti-CA12 Mouse Monoclonal Antibody [clone: 2E9]

Dodavatel: OriGene
ORIGCF505570EA 21120 CZK
ORIGCF505570
Anti-CA12 Mouse Monoclonal Antibody [clone: 2E9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CA12
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR