Tisk…

Anti-RHOJ Mouse Monoclonal Antibody [clone: 5C7]

Dodavatel: OriGene
ORIGCF505594EA 21120 CZK
ORIGCF505594
Anti-RHOJ Mouse Monoclonal Antibody [clone: 5C7]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: RHOJ
Clonality: Monoclonal
Clone: 5C7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR