Tisk…

Anti-RHOJ Mouse Monoclonal Antibody [clone: 5E5]

Dodavatel: OriGene
ORIGCF505595EA 21120 CZK
ORIGCF505595
Anti-RHOJ Mouse Monoclonal Antibody [clone: 5E5]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RHOJ
Clonality: Monoclonal
Clone: 5E5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR