Tisk…

Anti-MIER2 Mouse Monoclonal Antibody [clone: 3A1]

Dodavatel: OriGene
ORIGCF504403EA 21120 CZK
ORIGCF504403
Anti-MIER2 Mouse Monoclonal Antibody [clone: 3A1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MIER2
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR