Tisk…

Anti-SPG7 Mouse Monoclonal Antibody [clone: 1D2]

Dodavatel: OriGene
ORIGCF504423EA 21120 CZK
ORIGCF504423
Anti-SPG7 Mouse Monoclonal Antibody [clone: 1D2]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SPG7
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR