Tisk…

Anti-MIF4GD Mouse Monoclonal Antibody [clone: 4C11]

Dodavatel: OriGene
ORIGCF504443EA 21120 CZK
ORIGCF504443
Anti-MIF4GD Mouse Monoclonal Antibody [clone: 4C11]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MIF4GD
Clonality: Monoclonal
Clone: 4C11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR