Tisk…

Anti-ASCC2 Mouse Monoclonal Antibody [clone: 2D3]

Dodavatel: OriGene
ORIGCF503246EA 21120 CZK
ORIGCF503246
Anti-ASCC2 Mouse Monoclonal Antibody [clone: 2D3]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: ASCC2
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR