Tisk…

Anti-CD44 Mouse Monoclonal Antibody [clone: 5B4]

Dodavatel: OriGene
ORIGCF506749EA 21120 CZK
ORIGCF506749
Anti-CD44 Mouse Monoclonal Antibody [clone: 5B4]
Protilátky
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 5B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR