Tisk…

Anti-BID Mouse Monoclonal Antibody [clone: 2B6]

Dodavatel: OriGene
ORIGCF506755EA 21710 CZK
ORIGCF506755
Anti-BID Mouse Monoclonal Antibody [clone: 2B6]
Protilátky
Recommended Dilutions: Western Blot: 1:400-4000

Type: Primary
Antigen: BID
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR