Tisk…

Anti-BID Mouse Monoclonal Antibody [clone: 2A2]

Dodavatel: OriGene
ORIGCF506764EA 21710 CZK
ORIGCF506764
Anti-BID Mouse Monoclonal Antibody [clone: 2A2]
Protilátky
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: BID
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR