Tisk…

Anti-MMP13 Mouse Monoclonal Antibody [clone: 2A6]

Dodavatel: OriGene
ORIGCF506803EA 21120 CZK
ORIGCF506803
Anti-MMP13 Mouse Monoclonal Antibody [clone: 2A6]
Protilátky
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MMP13
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR