Tisk…

Anti-ETV7 Mouse Monoclonal Antibody [clone: 3B2]

Dodavatel: OriGene
ORIGCF802936EA 21120 CZK
ORIGCF802936
Anti-ETV7 Mouse Monoclonal Antibody [clone: 3B2]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ETV7
Clonality: Monoclonal
Clone: 3B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR