Tisk…

Anti-CD68 Mouse Monoclonal Antibody [clone: 3B6]

Dodavatel: OriGene
ORIGCF802949EA 21120 CZK
ORIGCF802949
Anti-CD68 Mouse Monoclonal Antibody [clone: 3B6]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 3B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR