Tisk…

Anti-C12orf26 Mouse Monoclonal Antibody [clone: 1E2]

Dodavatel: OriGene
ORIGCF507302EA 21710 CZK
ORIGCF507302
Anti-C12orf26 Mouse Monoclonal Antibody [clone: 1E2]
Protilátky
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: C12orf26
Clonality: Monoclonal
Clone: 1E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR