Tisk…

Anti-ANKMY2 Mouse Monoclonal Antibody [clone: 2G1]

Dodavatel: OriGene
ORIGCF507307EA 21710 CZK
ORIGCF507307
Anti-ANKMY2 Mouse Monoclonal Antibody [clone: 2G1]
Protilátky
Recommended Dilutions: Western Blot: 1:200-1000

Type: Primary
Antigen: ANKMY2
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR