Tisk…

Anti-SPN Mouse Monoclonal Antibody [clone: 2C7]

Dodavatel: OriGene
ORIGCF800840EA 21120 CZK
ORIGCF800840
Anti-SPN Mouse Monoclonal Antibody [clone: 2C7]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SPN
Clonality: Monoclonal
Clone: 2C7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR