Tisk…

Anti-CD99 Mouse Monoclonal Antibody [clone: 5C8]

Dodavatel: OriGene
ORIGCF800904EA 21120 CZK
ORIGCF800904
Anti-CD99 Mouse Monoclonal Antibody [clone: 5C8]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: 5C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR