Tisk…

Anti-PTH Mouse Monoclonal Antibody [clone: 1B10]

Dodavatel: OriGene
ORIGCF800912EA 21710 CZK
ORIGCF800912
Anti-PTH Mouse Monoclonal Antibody [clone: 1B10]
Protilátky
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: PTH
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR