Tisk…

Anti-PIK3C2A Mouse Monoclonal Antibody [clone: 3H2]

Dodavatel: OriGene
ORIGCF801692EA 21120 CZK
ORIGCF801692
Anti-PIK3C2A Mouse Monoclonal Antibody [clone: 3H2]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PIK3C2A
Clonality: Monoclonal
Clone: 3H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR