Tisk…

Anti-PIK3C2A Mouse Monoclonal Antibody [clone: 2F10]

Dodavatel: OriGene
ORIGCF801709EA 21120 CZK
ORIGCF801709
Anti-PIK3C2A Mouse Monoclonal Antibody [clone: 2F10]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PIK3C2A
Clonality: Monoclonal
Clone: 2F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR