Tisk…

Anti-FGFR1 Mouse Monoclonal Antibody [clone: 1A3]

Dodavatel: OriGene
ORIGCF802990EA 21120 CZK
ORIGCF802990
Anti-FGFR1 Mouse Monoclonal Antibody [clone: 1A3]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FGFR1
Clonality: Monoclonal
Clone: 1A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR