Tisk…

Anti-MRPL13 Mouse Monoclonal Antibody [clone: 6A11]

Dodavatel: OriGene
ORIGCF803030EA 21120 CZK
ORIGCF803030
Anti-MRPL13 Mouse Monoclonal Antibody [clone: 6A11]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MRPL13
Clonality: Monoclonal
Clone: 6A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR