Tisk…

Anti-PLK1 Rabbit Polyclonal Antibody

Dodavatel: OriGene
ORIGTA319250EA 24250 CZK
ORIGTA319250
Anti-PLK1 Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: ELISA: 1:3000-1:12000; Western Blot: 1:200-1:2000; Immunochemistry: 1:200-1:1000

Type: Primary
Antigen: PLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR