|
|
OBJEDNÁVKA
Produktová specifikace
Symbol antigenu | CCL20 |
Reaktivita | Human |
Spojení | Unconjugated |
Klonovalita | Monoclonal |
Druh protilátky | Primary |
Klon | 3N2D9 |
Hostitel | Mouse |
Western blot | Yes |
Epitop | full length |
Křížová adsorbce | No |
Imunogen | Anti-MIP-3a (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with mature full length recombinant human MIP-3a produced in E.coli followed by hybridoma development. |
Forma | Liquid |
Skladovací pufr | 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2 |
Skladovací teplota | – 20 °C |
Transportní teplota | –20 °C |