Tisk…

Anti-Esrp2 Mouse Monoclonal Antibody [clone: 22C5.H7.A6]

Dodavatel: OriGene
ORIGTA319604EA 24720 CZK
ORIGTA319604
Anti-Esrp2 Mouse Monoclonal Antibody [clone: 22C5.H7.A6]
Protilátky
Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:1000

Type: Primary
Antigen: Esrp2
Clonality: Monoclonal
Clone: 22C5.H7.A6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype:
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR