Tisk…

Anti-CD160 Rat Polyclonal Antibody [clone: eBioCNX46-3 (CNX46-3)]

Dodavatel: OriGene
ORIGTA320237EA 7450 CZK
ORIGTA320237
Anti-CD160 Rat Polyclonal Antibody [clone: eBioCNX46-3 (CNX46-3)]
Protilátky
Type: Primary
Antigen: CD160
Clonality: Polyclonal
Clone: eBioCNX46-3 (CNX46-3)
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR