Tisk…

Anti-CD180 Mouse Polyclonal Antibody [clone: MHR73-11]

Dodavatel: OriGene
ORIGTA320239EA 7840 CZK
ORIGTA320239
Anti-CD180 Mouse Polyclonal Antibody [clone: MHR73-11]
Protilátky
Type: Primary
Antigen: CD180
Clonality: Polyclonal
Clone: MHR73-11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR