Tisk…

Anti-CD1D Mouse Polyclonal Antibody [clone: 51.1]

Dodavatel: OriGene
ORIGTA320245EA 9020 CZK
ORIGTA320245
Anti-CD1D Mouse Polyclonal Antibody [clone: 51.1]
Protilátky
Type: Primary
Antigen: CD1D
Clonality: Polyclonal
Clone: 51.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR