Tisk…

Anti-CD27 Mouse Polyclonal Antibody [clone: O323]

Dodavatel: OriGene
ORIGTA320253EA 8570 CZK
ORIGTA320253
Anti-CD27 Mouse Polyclonal Antibody [clone: O323]
Protilátky
Type: Primary
Antigen: CD27
Clonality: Polyclonal
Clone: O323
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR