Tisk…

Anti-CD28 Golden Syrian Hamster Monoclonal Antibody [clone: 37.51]

Dodavatel: OriGene
ORIGTA320257EA 8570 CZK
ORIGTA320257
Anti-CD28 Golden Syrian Hamster Monoclonal Antibody [clone: 37.51]
Protilátky
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 37.51
Conjugation: Unconjugated
Epitope:
Host: Golden Syrian Hamster
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR