Tisk…

Anti-CD28 Mouse Polyclonal Antibody [clone: CD28.2]

Dodavatel: OriGene
ORIGTA320258EA 8260 CZK
ORIGTA320258
Anti-CD28 Mouse Polyclonal Antibody [clone: CD28.2]
Protilátky
Type: Primary
Antigen: CD28
Clonality: Polyclonal
Clone: CD28.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR