Tisk…

Anti-CD28 Mouse Polyclonal Antibody [clone: JJ319]

Dodavatel: OriGene
ORIGTA320259EA 16270 CZK
ORIGTA320259
Anti-CD28 Mouse Polyclonal Antibody [clone: JJ319]
Protilátky
Type: Primary
Antigen: CD28
Clonality: Polyclonal
Clone: JJ319
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR