Tisk…

Anti-CD38 Rat Polyclonal Antibody [clone: 90]

Dodavatel: OriGene
ORIGTA320266EA 8740 CZK
ORIGTA320266
Anti-CD38 Rat Polyclonal Antibody [clone: 90]
Protilátky
Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone: 90
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR