Tisk…

Anti-IgY Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Agrisera
AGRIAS09603EA 6150 CZK
AGRIAS09603
Anti-IgY Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR