Tisk…

Anti-IgG (H+L) Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: Agrisera
AGRIAS101179EA 6380 CZK
AGRIAS101179
Anti-IgG (H+L) Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Donkey
Isotype:
Reactivity: Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit, Rat, Sheep
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR