Tisk…

Anti-IgG (H+L) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Agrisera
AGRIAS101420EA 6940 CZK
AGRIAS101420
Anti-IgG (H+L) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Llama
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR