Tisk…

Anti-Biotin Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: Agrisera
AGRIAS101452EA 6570 CZK
AGRIAS101452
Anti-Biotin Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Type: Secondary
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR