Tisk…

Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)

Dodavatel: Agrisera
AGRIAS10658EA 5500 CZK
AGRIAS10658
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Guinea Pig
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR