Tisk…

Anti-IgG F(c) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: Agrisera
AGRIAS10813EA 7330 CZK
AGRIAS10813
Anti-IgG F(c) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR