Tisk…

Anti-IgM µ Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: Agrisera
AGRIAS10826EA 6210 CZK
AGRIAS10826
Anti-IgM µ Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR